Assay Details
| Gene Symbol: Tsg101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021884.3 | NP_068684.1 | 8 - 9 | 911 | 108 | |
| GenBank mRNA | AK014049.1 | - | 8 - 9 | 869 | 108 | |
| AK146229.1 | - | 8 - 9 | 921 | 108 | ||
| AK146602.1 | - | 8 - 9 | 912 | 108 | ||
| AK150369.1 | - | 8 - 9 | 950 | 108 | ||
| AK205423.1 | - | 1 - 2 | 120 | 109 | ||
| BC005424.1 | - | 8 - 9 | 927 | 3490301 | 108 | |
| BC085308.1 | - | 8 - 9 | 931 | 6395694 | 108 | |
| U52945.1 | - | 8 - 9 | 875 | 108 | ||
| X82319.1 | - | 1 - 2 | 122 | 108 | ||
More Information
Set Membership: |
|

Tsg101
Genomic Map