Assay Details
| Gene Symbol: Mir6925 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105890.1 | 1 | 33 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | kinesin family member 1C | Kif1c |
B430105J22Rik, D11Bwg1349e, Ltxs1 |
Sense | Chr. 11 - 70700548 - 70731970 on NCBI Build GRCm38 | |
| Downstream | predicted gene 12320 | Gm12320 | OTTMUSG00000006074 | 42.7kb | Anti-sense | Chr. 11 - 70748710 - 70755325 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Mir6925
Genomic Map