Assay Details
| Gene Symbol: Gm11681 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BE865132.1 | - | 1 - 2 | 123 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SRY (sex determining region Y)-box 9 | Sox9 |
2010306G03Rik, AV220920, mKIAA4243 |
-5.4kb | Anti-sense | Chr. 11 - 112782210 - 112787757 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700012H19 gene | 1700012H19Rik | 24.1kb | Sense | Chr. 11 - 112822965 - 112825128 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm11681
Genomic Map