Assay Details
| Gene Symbol: 1700024N05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006308.1 | - | 3 - 4 | 366 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
olfactory receptor 237, pseudogene 1 |
Olfr237-ps1 |
MOR261-4, Olfr438 |
-8.2kb | Anti-sense | Chr. 6 - 43153307 - 43154239 on NCBI Build GRCm38 |
| Downstream | olfactory receptor 437 | Olfr437 | MOR261-11 | 3.3kb | Anti-sense | Chr. 6 - 43167060 - 43167992 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

1700024N05Rik
Genomic Map