Assay Details
| Gene Symbol: LOC101055749 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX636598.1 | - | 1 - 2 | 239 | 147 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 44 | Gm44 | -262.0kb | Anti-sense | Chr. X - 90878355 - 90893134 on NCBI Build GRCm38 | |
| Downstream |
melanoma antigen, family B, 2 |
Mageb2 |
Mage-b2, Mage-rs2, Smage2 |
176.1kb | Anti-sense | Chr. X - 91331905 - 91332897 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

LOC101055749
Genomic Map