Assay Details
| Gene Symbol: Gm10371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK156967.1 | - | 1 - 2 | 56 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sterile alpha motif domain containing 4 | Samd4 |
1700024G08Rik, 1700111L17Rik, 4933436G17Rik, Samd4a, Smaug, Smaug1 |
-9.4kb | Anti-sense | Chr. 14 - 46882965 - 47105817 on NCBI Build GRCm38 |
| Downstream | GTP cyclohydrolase 1 | Gch1 |
GTP-CH, GTPCH, Gch |
37.7kb | Sense | Chr. 14 - 47153895 - 47189402 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK156967.1 transcripts.
Set Membership: |
|

Gm10371
Genomic Map