Assay Details
| Gene Symbol: 4833422M21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014752.1 | - | 1 - 2 | 70 | ||
| AK084200.1 | - | 1 - 2 | 224 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
eukaryotic translation initiation factor 3, subunit I pseudogene |
Gm4371 |
EG667874, Gm8854 |
-50.4kb | Anti-sense | Chr. 15 - 96182800 - 96203470 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA E330033B04 gene | E330033B04Rik | 13.9kb | Sense | Chr. 15 - 96268564 - 96275275 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4833422M21Rik
Genomic Map