Assay Details
| Gene Symbol: Gm11665 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK139889.1 | - | 5 - 6 | 913 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cell division cycle 27 | Cdc27 |
AI452358, APC3, BC023187 |
-6.2kb | Sense | Chr. 11 - 104502524 - 104550620 on NCBI Build GRCm38 |
| Downstream | integrin beta 3 | Itgb3 |
CD61, GP3A, INGRB3 |
50.2kb | Anti-sense | Chr. 11 - 104608000 - 104670490 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm11665
Genomic Map