Assay Details
| Gene Symbol: 1700123J17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007252.1 | - | 3 - 4 | 198 | 82 | |
| AK043947.1 | - | 4 - 5 | 372 | 82 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | exportin 6 | Xpo6 |
2610005L19Rik, AI854665, AL022631, C230091E20Rik, R75304, Ranbp20, mKIAA0370 |
-24.9kb | Sense | Chr. 7 - 126101718 - 126200496 on NCBI Build GRCm38 |
| Downstream | SH3-binding kinase 1 | Sbk1 | Sbk | 45.4kb | Anti-sense | Chr. 7 - 126272619 - 126294999 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

1700123J17Rik
Genomic Map