Assay Details
| Gene Symbol: Bloodlinc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BB629591.1 | - | 2 - 3 | 337 | 110 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 4 (anion exchanger), member 1 |
Slc4a1 |
Ae1, CD233, Empb3, l11Jus51 |
-10.3kb | Sense | Chr. 11 - 102348820 - 102365281 on NCBI Build GRCm38 |
| Downstream | RUN domain containing 3A | Rundc3a |
RPIP-8, Rap2ip, Rpip8 |
17.0kb | Anti-sense | Chr. 11 - 102393403 - 102402939 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Bloodlinc
Genomic Map