Assay Details
| Gene Symbol: Gm11729 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AI854885.1 | - | 1 - 2 | 200 | 119 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | septin 9 | Sept9 |
MSF1, Msf, PNUTL4, Sint1 |
-52.5kb | Anti-sense | Chr. 11 - 117199661 - 117362325 on NCBI Build GRCm38 |
| Downstream | predicted gene 11732 | Gm11732 | OTTMUSG00000003800 | 10.8kb | Anti-sense | Chr. 11 - 117426163 - 117438866 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm11729
Genomic Map