Assay Details
| Gene Symbol: LOC102640359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC094633.1 | - | 1 - 2 | 148 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6992 | Mir6992 |
Gm27906, mmu-mir-6992 |
-11.7kb | Anti-sense | Chr. 19 - 8742971 - 8743079 on NCBI Build GRCm38 |
| Downstream | nuclear RNA export factor 1 | Nxf1 |
Mex67, Mvb1, Tap |
178.0 | Anti-sense | Chr. 19 - 8757103 - 8770910 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

LOC102640359
Genomic Map