Assay Details
| Gene Symbol: Gm29669 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK079925.1 | - | 268 | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | protein tyrosine phosphatase 4a1 | Ptp4a1 |
AA415290, AU019864, C130021B01, Prl-1 |
-71.1kb | Sense | Chr. 1 - 30940303 - 30949765 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm29669
Genomic Map