Assay Details
| Gene Symbol: Gm12863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK142602.1 | - | 1 - 2 | 119 | 100 | |
| BB506114.2 | - | 1 - 2 | 129 | 100 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | olfactory receptor 1328 | Olfr1328 |
MOR259-1, MOR259-13, Olfr1519 |
-19.4kb | Sense | Chr. 4 - 118933899 - 118934840 on NCBI Build GRCm38 |
| Downstream | L-amino acid oxidase 1 | Lao1 | AW990848 | 7.0kb | Anti-sense | Chr. 4 - 118961967 - 118968910 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK142602.1 transcripts.
Set Membership: |
|

Gm12863
Genomic Map