Assay Details
| Gene Symbol: Nudt12os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK132847.1 | - | 1 - 2 | 242 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PDZ and pleckstrin homology domains 1 | Pdzph1 |
2610034M16Rik, AV114557 |
-4.9kb | Anti-sense | Chr. 17 - 58878808 - 58991375 on NCBI Build GRCm38 |
| Downstream | ephrin A5 | Efna5 |
AL-1, AV158822, EFL-5, Ephrin-A5, Epl7, LERK-7, RAGS |
3.6mb | Anti-sense | Chr. 17 - 62602957 - 62881317 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK132847.1 transcripts.
Set Membership: |
|

Nudt12os
Genomic Map