Assay Details
| Gene Symbol: Gm12300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK009974.1 | - | 1 - 2 | 270 | 137 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
myosin, heavy polypeptide 2, skeletal muscle, adult |
Myh2 |
MHC2A, MyHC-IIa, Myh2a, Myhs-f, Myhs-f1, Myhsf1 |
-26.7kb | Anti-sense | Chr. 11 - 67171027 - 67197517 on NCBI Build GRCm38 |
| Downstream |
myosin, heavy polypeptide 4, skeletal muscle |
Myh4 |
AI506973, MHC2B, MM, MYH-2B, Minimsc, Minmus, MyHC-IIb, Myhsf |
531.0 | Anti-sense | Chr. 11 - 67237812 - 67260447 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm12300
Genomic Map