Assay Details
| Gene Symbol: BB453382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK083850.1 | - | 1 | 2925 | 97 | |
| BB453382.2 | - | 1 | 546 | 97 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BB453382| Gene Symbol: BB453382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK083850.1 | - | 1 | 2925 | 97 | |
| BB453382.2 | - | 1 | 546 | 97 | ||
Set Membership: |
|