Assay Details
| Gene Symbol: Zfp706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026521.4 | NP_080797.1 | 4 | 2303 | 128 | |
| GenBank mRNA | AB041652.1 | - | 4 | 2246 | 128 | |
| AK004003.1 | - | 1 | 695 | 128 | ||
| AK032231.1 | - | 4 | 2189 | 128 | ||
| AK082392.1 | - | 4 | 2207 | 128 | ||
| AK132053.1 | - | 4 | 2220 | 128 | ||
| AK145319.1 | - | 4 | 2181 | 128 | ||
| AK166530.1 | - | 4 | 2181 | 128 | ||
| AK167897.1 | - | 4 | 2220 | 128 | ||
| AK168958.1 | - | 4 | 2182 | 128 | ||
| BC037600.1 | - | 4 | 2228 | 128 | ||
| BC042704.1 | - | 4 | 2194 | 128 | ||
| BC052459.1 | - | 4 | 2303 | 128 | ||
| BC054356.1 | - | 4 | 2225 | 128 | ||
| BC085477.1 | - | 4 | 2187 | 128 | ||
| BC147306.1 | - | 4 | 2162 | 9056379 | 128 | |
| BC147307.1 | - | 4 | 2162 | 9056367 | 128 | |
More Information
Set Membership: |
|

Zfp706
Genomic Map