Assay Details
| Gene Symbol: Cttnbp2nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163332.1 | NP_001156804.1 | 6 | 2612 | 85 | |
| NM_001163333.1 | NP_001156805.1 | 6 | 2604 | 85 | ||
| NM_030249.4 | NP_084525.1 | 6 | 2720 | 85 | ||
| GenBank mRNA | AK029901.1 | - | 6 | 2707 | 85 | |
| AK030438.1 | - | 6 | 2585 | 85 | ||
| AK129359.1 | - | 6 | 2610 | 85 | ||
| AK145085.1 | - | 6 | 2586 | 85 | ||
| BC003236.1 | - | 6 | 2574 | 3498078 | 85 | |
| BC006952.1 | - | 2 | 2062 | 85 | ||
More Information
Set Membership: |
|

Cttnbp2nl
Genomic Map