Assay Details
| Gene Symbol: Sbspon | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK140569.1 | - | 2516 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sbspon| Gene Symbol: Sbspon | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK140569.1 | - | 2516 | 94 | ||