Assay Details
| Gene Symbol: Mcm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008563.2 | NP_032589.1 | 17 | 2770 | 121 | |
| GenBank mRNA | AK088142.1 | - | 16 | 3275 | 121 | |
| AK133850.1 | - | 17 | 2835 | 121 | ||
| AK145563.1 | - | 17 | 2869 | 121 | ||
| AK146497.1 | - | 17 | 2783 | 121 | ||
| AK147065.1 | - | 17 | 2785 | 121 | ||
| AK148006.1 | - | 17 | 2815 | 121 | ||
| BC020400.1 | - | 17 | 2784 | 121 | ||
| BC031700.1 | - | 17 | 2769 | 5007886 | 121 | |
More Information
Set Membership: |
|

Mcm3
Genomic Map