Assay Details
| Gene Symbol: Yae1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025904.3 | NP_080180.1 | 3 | 457 | 148 | |
| GenBank mRNA | AK005425.1 | - | 3 | 444 | 148 | |
| AK011774.1 | - | 3 | 456 | 148 | ||
| AK011805.1 | - | 3 | 425 | 148 | ||
| AK011852.1 | - | 3 | 425 | 148 | ||
| AK019184.1 | - | 3 | 411 | 148 | ||
| AK076034.1 | - | 3 | 414 | 148 | ||
| AK151981.1 | - | 3 | 435 | 148 | ||
| BC020123.1 | - | 3 | 148 | |||
| BC037189.1 | - | 3 | 148 | |||
| BC048453.1 | - | 3 | 440 | 6595060 | 148 | |
| BC083111.1 | - | 3 | 5329860 | 148 | ||
More Information
Set Membership: |
|

Yae1d1
Genomic Map