Assay Details
| Gene Symbol: Supt20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019995.3 | NP_064379.1 | 15 - 16 | 1368 | 61 | |
| GenBank mRNA | AF139179.1 | - | 15 - 16 | 1372 | 61 | |
| BC052702.1 | - | 15 - 16 | 1357 | 6400922 | 61 | |
More Information
Set Membership: |
|

Supt20
Genomic Map