Assay Details
| Gene Symbol: BB187690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK138502.1 | - | 1 | 613 | 95 | |
| AK138626.1 | - | 1 | 613 | 95 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BB187690| Gene Symbol: BB187690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK138502.1 | - | 1 | 613 | 95 | |
| AK138626.1 | - | 1 | 613 | 95 | ||
Set Membership: |
|