Assay Details
| Gene Symbol: Pou3f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008899.2 | NP_032925.1 | 1 | 87 | ||
| GenBank mRNA | AK139058.1 | - | 1 | 125 | 87 | |
| X66602.1 | - | 1 | 87 | |||
More Information
Set Membership: |
|

Pou3f2
Genomic Map