Assay Details
| Gene Symbol: Megf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK155926.1 | - | 3319 | 97 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Megf9| Gene Symbol: Megf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK155926.1 | - | 3319 | 97 | ||