Assay Details
| Gene Symbol: Sh3gl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK047324.1 | - | 3579 | 96 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh3gl2| Gene Symbol: Sh3gl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK047324.1 | - | 3579 | 96 | ||