Assay Details
| Gene Symbol: Grb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008163.3 | NP_032189.1 | 2 - 3 | 411 | 106 | |
| GenBank mRNA | AK077750.1 | - | 1 - 2 | 230 | 106 | |
| AK079087.1 | - | 1 - 2 | 238 | 106 | ||
| AK148148.1 | - | 2 - 3 | 376 | 106 | ||
| AK153557.1 | - | 1 - 2 | 344 | 106 | ||
| AK155800.1 | - | 2 - 3 | 366 | 106 | ||
| AK166142.1 | - | 2 - 3 | 365 | 106 | ||
| AK170845.1 | - | 1 - 2 | 202 | 106 | ||
| AK172002.1 | - | 2 - 3 | 386 | 106 | ||
| AK196759.1 | - | 2 - 3 | 235 | 106 | ||
| BC052377.1 | - | 2 - 3 | 411 | 5696248 | 106 | |
| BC085254.1 | - | 2 - 3 | 247 | 5343274 | 106 | |
| D85748.1 | - | 1 - 2 | 158 | 106 | ||
| U07617.1 | - | 1 - 2 | 160 | 106 | ||
More Information
Set Membership: |
|

Grb2
Genomic Map