Assay Details
| Gene Symbol: Szrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025608.2 | NP_001020779.1 | 4 | 1291 | 87 | |
| NM_001277195.1 | NP_001264124.1 | 4 | 1288 | 87 | ||
| NR_102343.1 | 3 | 1241 | 87 | |||
| NR_102344.1 | 3 | 1238 | 87 | |||
| GenBank mRNA | AK145912.1 | - | 3 | 87 | ||
| AK171702.1 | - | 3 | 1214 | 87 | ||
| AK184848.1 | - | 1 | 100 | 87 | ||
| BC066992.1 | - | 4 | 1239 | 5685985 | 87 | |
More Information
Set Membership: |
|

Szrd1
Genomic Map