Assay Details
| Gene Symbol: Hnrnph2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019868.3 | NP_063921.1 | 4 | 2200 | 141 | |
| GenBank mRNA | AK077716.1 | - | 4 | 2201 | 141 | |
| AK168116.1 | - | 3 | 2183 | 141 | ||
| AK169104.1 | - | 3 | 2166 | 142 | ||
| BC005461.1 | - | 3 | 2182 | 3496629 | 141 | |
More Information
Set Membership: |
|

Hnrnph2
Genomic Map