Assay Details
| Gene Symbol: Kdm2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013910.2 | NP_038938.1 | 10 - 11 | 2086 | 95 | |
| GenBank mRNA | AF176524.2 | - | 3 - 4 | 507 | 95 | |
| AK031626.1 | - | 10 - 11 | 2052 | 95 | ||
| AK034033.1 | - | 4 - 5 | 1080 | 95 | ||
| AK129479.1 | - | 21 - 22 | 3523 | 95 | ||
| BC057622.1 | - | 10 - 11 | 2081 | 6402440 | 95 | |
| BC065090.1 | - | 21 - 22 | 3722 | 6851421 | 95 | |
More Information
Set Membership: |
|

Kdm2b
Genomic Map