Assay Details
| Gene Symbol: Eef2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007907.2 | NP_031933.1 | 11 | 2046 | 83 | |
| GenBank mRNA | AK028938.1 | - | 11 | 2025 | 83 | |
| AK032997.1 | - | 11 | 2032 | 83 | ||
| AK040474.1 | - | 11 | 2032 | 83 | ||
| AK077866.1 | - | 11 | 2032 | 83 | ||
| AK087985.1 | - | 11 | 2032 | 83 | ||
| AK128976.1 | - | 11 | 2031 | 83 | ||
| AK134195.1 | - | 11 | 2032 | 83 | ||
| AK144874.1 | - | 11 | 2034 | 83 | ||
| AK145509.1 | - | 11 | 2033 | 83 | ||
| AK145545.1 | - | 11 | 2032 | 83 | ||
| AK146508.1 | - | 11 | 2034 | 83 | ||
| AK146947.1 | - | 11 | 2032 | 83 | ||
| AK149659.1 | - | 11 | 2034 | 84 | ||
| AK150136.1 | - | 11 | 2032 | 83 | ||
| AK150902.1 | - | 11 | 2032 | 83 | ||
| AK151487.1 | - | 11 | 2032 | 83 | ||
| AK151812.1 | - | 11 | 1982 | 83 | ||
| AK151945.1 | - | 11 | 2032 | 83 | ||
| AK152146.1 | - | 11 | 2032 | 83 | ||
| AK152447.1 | - | 11 | 2032 | 83 | ||
| AK152587.1 | - | 11 | 2032 | 83 | ||
| AK152826.1 | - | 11 | 2032 | 83 | ||
| AK153275.1 | - | 11 | 2032 | 83 | ||
| AK155325.1 | - | 11 | 2032 | 83 | ||
| AK159155.1 | - | 11 | 2033 | 83 | ||
| AK159421.1 | - | 11 | 2032 | 83 | ||
| AK159514.1 | - | 11 | 2032 | 83 | ||
| AK159806.1 | - | 11 | 2032 | 83 | ||
| AK161993.1 | - | 4 | 632 | 83 | ||
| AK163063.1 | - | 11 | 2032 | 83 | ||
| AK164406.1 | - | 3 | 478 | 83 | ||
| AK166212.1 | - | 11 | 2032 | 83 | ||
| AK166596.1 | - | 11 | 2032 | 83 | ||
| AK166719.1 | - | 11 | 2034 | 83 | ||
| AK166747.1 | - | 11 | 2032 | 83 | ||
| AK166751.1 | - | 11 | 2030 | 83 | ||
| AK166758.1 | - | 11 | 2033 | 83 | ||
| AK166788.1 | - | 11 | 2034 | 83 | ||
| AK166815.1 | - | 11 | 2033 | 83 | ||
| AK166829.1 | - | 11 | 2033 | 83 | ||
| AK166851.1 | - | 11 | 2030 | 83 | ||
| AK166968.1 | - | 11 | 2032 | 83 | ||
| AK167081.1 | - | 11 | 2033 | 83 | ||
| AK167093.1 | - | 11 | 2032 | 83 | ||
| AK167109.1 | - | 11 | 2032 | 83 | ||
| AK167115.1 | - | 11 | 2032 | 83 | ||
| AK167199.1 | - | 11 | 2032 | 83 | ||
| AK167221.1 | - | 11 | 2032 | 83 | ||
| AK167281.1 | - | 11 | 2032 | 83 | ||
| AK168527.1 | - | 11 | 2033 | 83 | ||
| AK168615.1 | - | 11 | 2033 | 83 | ||
| AK168638.1 | - | 11 | 2033 | 83 | ||
| AK168659.1 | - | 11 | 2033 | 83 | ||
| AK168794.1 | - | 11 | 2032 | 83 | ||
| AK168827.1 | - | 11 | 2032 | 83 | ||
| AK168874.1 | - | 11 | 2025 | 83 | ||
| AK169013.1 | - | 11 | 2032 | 83 | ||
| BC002233.1 | - | 1 | 238 | 83 | ||
| BC007152.1 | - | 11 | 2030 | 3600352 | 83 | |
| BC013263.1 | - | 11 | 2018 | 83 | ||
| BC060707.1 | - | 10 | 1907 | 83 | ||
| M76131.1 | - | 1 | 180 | 83 | ||
More Information
Set Membership: |
|

Eef2
Genomic Map