Assay Details
| Gene Symbol: Ywhag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018871.3 | NP_061359.2 | 2 | 1993 | 125 | |
| GenBank mRNA | AK088847.1 | - | 2 | 1907 | 125 | |
| AK153307.1 | - | 2 | 1950 | 125 | ||
| AK164356.1 | - | 2 | 1944 | 125 | ||
| BC008129.1 | - | 2 | 1876 | 3498633 | 125 | |
More Information
Set Membership: |
|

Ywhag
Genomic Map