Assay Details
| Gene Symbol: BB187676 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK046110.1 | - | 1 | 518 | 81 | |
| BB313560.2 | - | 1 | 505 | 81 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BB187676| Gene Symbol: BB187676 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK046110.1 | - | 1 | 518 | 81 | |
| BB313560.2 | - | 1 | 505 | 81 | ||
Set Membership: |
|