Assay Details
| Gene Symbol: Syn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013681.3 | NP_038709.1 | 11 | 76 | ||
| GenBank mRNA | AK013786.1 | - | 1 | 619 | 76 | |
| AK029181.1 | - | 9 | 76 | |||
More Information
Set Membership: |
|

Syn2
Genomic Map