Assay Details
| Gene Symbol: Uck1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011675.2 | NP_035805.2 | 7 | 847 | 66 | |
| GenBank mRNA | AK132100.1 | - | 2 | 119 | 66 | |
| BC025146.1 | - | 7 | 814 | 4913412 | 66 | |
| L31783.1 | - | 7 | 707 | 66 | ||
More Information
Set Membership: |
|

Uck1
Genomic Map