Assay Details
| Gene Symbol: 6430511E19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK032249.1 | - | 1 | 827 | 86 | |
| AK081851.1 | - | 1 | 86 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

6430511E19Rik| Gene Symbol: 6430511E19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK032249.1 | - | 1 | 827 | 86 | |
| AK081851.1 | - | 1 | 86 | |||
Set Membership: |
|