Assay Details
| Gene Symbol: Prkcdbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK048894.1 | - | 723 | 82 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkcdbp| Gene Symbol: Prkcdbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK048894.1 | - | 723 | 82 | ||