Assay Details
| Gene Symbol: Tpm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK049383.1 | - | 1 | 2019 | 83 | |
| AK171835.1 | - | 1 | 1427 | 83 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Tpm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK049383.1 | - | 1 | 2019 | 83 | |
| AK171835.1 | - | 1 | 1427 | 83 | ||
Set Membership: |
|