Assay Details
| Gene Symbol: Pinc | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003202.1 | 5 | 584 | 84 | |||
| GenBank mRNA | DQ059755.1 | - | LIT3460 PINC |
5 | 584 | 84 | |
More Information
Set Membership: |
|

Pinc
Genomic Map