Assay Details
| Gene Symbol: Lsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026032.1 | NP_080308.1 | 4 | 599 | 125 | |
| GenBank mRNA | AK006006.1 | - | 2 | 334 | 125 | |
| AK012814.1 | - | 2 | 334 | 125 | ||
| AK019398.1 | - | 2 | 329 | 125 | ||
| AK041290.1 | - | 4 | 599 | 125 | ||
| BC021460.1 | - | 4 | 882 | 5054997 | 125 | |
More Information
Set Membership: |
|

Lsm1
Genomic Map