Assay Details
| Gene Symbol: Sh3rf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021506.2 | NP_067481.2 | 12 | 106 | ||
| GenBank mRNA | AK008634.1 | - | 1 | 124 | 106 | |
| AK136410.1 | - | 1 | 106 | |||
| BC060113.1 | - | 13 | 6405370 | 106 | ||
| BC060696.1 | - | 13 | 6405370 | 106 | ||
| CO040875.1 | - | 1 | 221 | 106 | ||
More Information
Set Membership: |
|

Sh3rf1
Genomic Map