Assay Details
| Gene Symbol: Cbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK152275.1 | - | 1 | 519 | 129 | |
| AK154208.1 | - | 1 | 129 | |||
| AK171171.1 | - | 1 | 129 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Cbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK152275.1 | - | 1 | 519 | 129 | |
| AK154208.1 | - | 1 | 129 | |||
| AK171171.1 | - | 1 | 129 | |||
Set Membership: |
|