Assay Details
Gene Symbol: Arcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK135317.1 | - | 1 | 822 | 105 | |
BC050874.1 | - | 1 | 1593 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Arcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK135317.1 | - | 1 | 822 | 105 | |
BC050874.1 | - | 1 | 1593 | 105 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |