Assay Details
| Gene Symbol: Ftx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028380.1 | 1 | 80 | 93 | ||
| NR_028381.1 | 1 | 80 | 93 | |||
| GenBank mRNA | AK035525.1 | - | 1 | 82 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Ftx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028380.1 | 1 | 80 | 93 | ||
| NR_028381.1 | 1 | 80 | 93 | |||
| GenBank mRNA | AK035525.1 | - | 1 | 82 | 93 | |
Set Membership: |
|