Assay Details
| Gene Symbol: Golga7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042484.1 | NP_001035949.1 | 6 | 1460 | 115 | |
| NM_020585.2 | NP_065610.2 | 5 | 1673 | 115 | ||
| GenBank mRNA | AB041568.1 | - | 5 | 1609 | 115 | |
| AK048175.1 | - | 6 | 1436 | 115 | ||
| AK077582.1 | - | 6 | 1434 | 115 | ||
| AK150462.1 | - | 6 | 1454 | 115 | ||
| AK150696.1 | - | 5 | 1615 | 115 | ||
| AK151353.1 | - | 6 | 1454 | 115 | ||
| AK152520.1 | - | 6 | 1454 | 115 | ||
| AK152606.1 | - | 6 | 1433 | 115 | ||
| AK153213.1 | - | 6 | 1454 | 115 | ||
| AK158227.1 | - | 5 | 1579 | 115 | ||
| AK170129.1 | - | 6 | 1439 | 115 | ||
| BC016894.1 | - | 6 | 1412 | 4223937 | 115 | |
| BC019697.1 | - | 5 | 1598 | 115 | ||
| BC058523.1 | - | 5 | 1516 | 5717934 | 115 | |
| BC093518.1 | - | 6 | 1430 | 3157520 | 115 | |
| BC103796.1 | - | 6 | 1425 | 6519018 | 115 | |
More Information
Set Membership: |
|

Golga7
Genomic Map