Assay Details
| Gene Symbol: Hnrnpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166427.1 | NP_001159899.1 | 3 | 1471 | 110 | |
| NM_001166428.1 | NP_001159900.1 | 4 | 1410 | 110 | ||
| NM_001166429.1 | NP_001159901.1 | 4 | 1371 | 110 | ||
| NM_001166430.1 | NP_001159902.1 | 4 | 1722 | 110 | ||
| NM_001166431.1 | NP_001159903.1 | 3 | 1661 | 110 | ||
| NM_001166432.1 | NP_001159904.1 | 4 | 1462 | 110 | ||
| NM_133834.2 | NP_598595.1 | 4 | 1353 | 110 | ||
| GenBank mRNA | AK076478.1 | - | 3 | 1311 | 110 | |
| BC018185.1 | - | 3 | 1296 | 3995067 | 110 | |
| BC023162.1 | - | 1 | 377 | 110 | ||
| BC023793.1 | - | 4 | 1348 | 110 | ||
| BC025465.1 | - | 1 | 87 | 110 | ||
| BC025481.1 | - | 4 | 1330 | 5354000 | 110 | |
| BC027003.1 | - | 1 | 930 | 5365491 | 110 | |
| BC029163.1 | - | 1 | 1114 | 4949890 | 110 | |
| BC029764.1 | - | 1 | 937 | 4947521 | 110 | |
| BC032197.1 | - | 4 | 1348 | 110 | ||
| BC033483.1 | - | 1 | 1112 | 4950131 | 110 | |
| BC042689.1 | - | 3 | 1594 | 110 | ||
| BC047346.1 | - | 3 | 1594 | 110 | ||
| BC089313.1 | - | 4 | 1195 | 6808998 | 110 | |
More Information
Set Membership: |
|

Hnrnpf
Genomic Map