Assay Details
| Gene Symbol: Pgrmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016783.3 | NP_058063.2 | 3 | 1715 | 123 | |
| GenBank mRNA | AF042491.1 | - | 3 | 1666 | 123 | |
| AK153272.1 | - | 3 | 1732 | 123 | ||
| AK159101.1 | - | 3 | 1732 | 123 | ||
| AK159345.1 | - | 3 | 1732 | 123 | ||
| AK159599.1 | - | 3 | 1732 | 123 | ||
| AK162315.1 | - | 3 | 1734 | 123 | ||
| AK166240.1 | - | 3 | 1732 | 123 | ||
| AK167655.1 | - | 3 | 1732 | 123 | ||
| AK168387.1 | - | 3 | 1731 | 123 | ||
| AK168882.1 | - | 3 | 1732 | 123 | ||
| AK169063.1 | - | 4 | 1675 | 123 | ||
| AK214839.1 | - | 1 | 286 | 123 | ||
| BC006016.1 | - | 3 | 1717 | 3591536 | 123 | |
More Information
Set Membership: |
|

Pgrmc1
Genomic Map