Assay Details
| Gene Symbol: 9230105E05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040626.1 | 3 | 969 | 114 | ||
| GenBank mRNA | AK033763.1 | - | 3 | 971 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

9230105E05Rik| Gene Symbol: 9230105E05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040626.1 | 3 | 969 | 114 | ||
| GenBank mRNA | AK033763.1 | - | 3 | 971 | 114 | |
Set Membership: |
|