Assay Details
| Gene Symbol: Usp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301628.1 | NP_001288557.1 | 21 | 6390 | 98 | |
| NM_027604.4 | NP_081880.2 | 22 | 6477 | 98 | ||
| GenBank mRNA | AK084098.1 | - | 1 | 1520 | 98 | |
More Information
Set Membership: |
|

Usp15
Genomic Map